ENAH5802E007 Enamine

5-(2,2,2-trifluoroethyl)piperidin-2-one

  • MDL number MFCD32065910
  • Cas Number : 2361645-50-1
  • Molecular Weight : 181.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(2,2,2-trifluoroethyl)piperidin-2-one
Inchi Code 1S/C7H10F3NO/c8-7(9,10)3-5-1-2-6(12)11-4-5/h5H,1-4H2,(H,11,12)
Inchi Key AZPZGXMWMXZVEO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430574
Availability details for
Quantity
  • Availability
  • Add to Cart