ENAH5802E00B Enamine

(3R,6R)-3-methyl-6-(2-methylpropyl)piperazine-2,5-dione

  • MDL number MFCD32197890
  • Cas Number : 5845-58-9
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 258-259
Iupac Name (3R,6R)-3-isobutyl-6-methylpiperazine-2,5-dione
Inchi Code 1S/C9H16N2O2/c1-5(2)4-7-9(13)10-6(3)8(12)11-7/h5-7H,4H2,1-3H3,(H,10,13)(H,11,12)/t6-,7-/m1/s1
Inchi Key DBJPZCJQDRPOME-RNFRBKRXSA-N

Description

Melting Point 258-259
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7441099

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7441099
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart