ENAH5802E014 Enamine

octahydro-2lambda6-thieno[3,4-c]pyridine-2,2,6-trione

  • MDL number MFCD32068660
  • Cas Number : 2375261-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434415
Availability details for
Quantity
  • Availability
  • Add to Cart