ENAH5802E015 Enamine

N-phenylcyclopentanecarboxamide

  • MDL number MFCD03362673
  • Cas Number : 3400-41-7
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431658
Availability details for
Quantity
  • Availability
  • Add to Cart