ENAH5802E016 Enamine

N-cyclopentylbenzamide

  • MDL number MFCD00996282
  • Cas Number : 53226-42-9
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-cyclopentylbenzamide
Inchi Code 1S/C12H15NO/c14-12(10-6-2-1-3-7-10)13-11-8-4-5-9-11/h1-3,6-7,11H,4-5,8-9H2,(H,13,14)
Inchi Key ZLDFSUZIEYDGHD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431622
Availability details for
Quantity
  • Availability
  • Add to Cart