ENAH5802E017 Enamine

methyl 2-oxo-1H,2H,3H-pyrrolo[2,3-c]pyridine-4-carboxylate

  • MDL number MFCD31706295
  • Cas Number : 2361643-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6732571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6732571
Availability details for
Quantity
  • Availability
  • Add to Cart