ENAH5802E018 Enamine

5-(furan-2-yl)-N-methyl-1,2-oxazole-3-carboxamide

  • MDL number MFCD19596632
  • Cas Number : 1343867-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194338
Availability details for
Quantity
  • Availability
  • Add to Cart