ENAH5802E01B Enamine

3-ethoxy-4-methoxybenzamide

  • MDL number MFCD10472295
  • Cas Number : 247569-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-197
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444384
Availability details for
Quantity
  • Availability
  • Add to Cart