ENAH5802E01F Enamine

3-(2,2-dimethylthiomorpholin-4-yl)propanamide

  • MDL number MFCD32068914
  • Cas Number : 1602346-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5332583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5332583
Availability details for
Quantity
  • Availability
  • Add to Cart