ENAH5802E021 Enamine

5-benzyl-3-methylpiperidin-2-one

  • MDL number MFCD24616292
  • Cas Number : 1239983-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5312781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5312781
Availability details for
Quantity
  • Availability
  • Add to Cart