ENAH5802E023 Enamine

N-ethyl-5-(furan-2-yl)-1,2-oxazole-3-carboxamide

  • MDL number MFCD19383565
  • Cas Number : 1461754-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194335
Availability details for
Quantity
  • Availability
  • Add to Cart