ENAH5802E024 Enamine

4-bromo-1,3-thiazole-2-carboxamide

  • MDL number MFCD12755855
  • Cas Number : 912639-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1700470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1700470
Availability details for
Quantity
  • Availability
  • Add to Cart