ENAH5802E028 Enamine

6-chloro-N-(cyclopropylmethyl)pyridine-3-carboxamide

  • MDL number MFCD09805007
  • Cas Number : 585544-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5029760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5029760
Availability details for
Quantity
  • Availability
  • Add to Cart