ENAH5802E02A Enamine

6-chloro-7-methoxy-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD24621773
  • Cas Number : 6501-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-177
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176663
Availability details for
Quantity
  • Availability
  • Add to Cart