ENAH5802E030 Enamine

5-(1,1,2,2,2-pentafluoroethyl)piperidin-2-one

  • MDL number MFCD32066271
  • Cas Number : 2361634-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434175
Availability details for
Quantity
  • Availability
  • Add to Cart