ENAH5802E032 Enamine

N-phenylcycloheptanecarboxamide

  • MDL number MFCD20048637
  • Cas Number : 18208-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431650
Availability details for
Quantity
  • Availability
  • Add to Cart