ENAH5802E033 Enamine

3,5-dichloro-N-ethylbenzamide

  • MDL number MFCD01325797
  • Cas Number : 33244-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437739
Availability details for
Quantity
  • Availability
  • Add to Cart