ENAH5802E036 Enamine

2-[2-oxo-2-(pyridin-3-yl)ethylidene]-1,3-thiazolidin-4-one

  • MDL number MFCD28155407
  • Cas Number : 1000932-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30654
Availability details for
Quantity
  • Availability
  • Add to Cart