ENAH5802E03A Enamine

2-amino-N-[3-(dimethylamino)propyl]benzamide

  • MDL number MFCD09186202
  • Cas Number : 1943-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7383194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7383194
Availability details for
Quantity
  • Availability
  • Add to Cart