ENAH5802E03D Enamine

6-chloro-N-(cyclobutylmethyl)pyridazine-3-carboxamide

  • MDL number MFCD12165865
  • Cas Number : 1179731-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5091805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5091805
Availability details for
Quantity
  • Availability
  • Add to Cart