ENAH5802E03F Enamine

6-chloro-N-(2,2-dimethylpropyl)pyridazine-3-carboxamide

  • MDL number MFCD14593589
  • Cas Number : 1250140-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433551

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433551
Availability details for
Quantity
  • Availability
  • Add to Cart