ENAH5802E041 Enamine

N-{5-chloro-[1,3]thiazolo[5,4-d]pyrimidin-2-yl}acetamide

  • Cas Number : 1204742-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442631
Availability details for
Quantity
  • Availability
  • Add to Cart