ENAH5802E046 Enamine

(3R)-7-bromo-3-methyl-1H,2H,3H,4H-pyrrolo[1,2-a]pyrazin-1-one

  • MDL number MFCD32066088
  • Cas Number : 1841360-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432753
Availability details for
Quantity
  • Availability
  • Add to Cart