ENAH5802E047 Enamine

7-(trifluoromethyl)-1,2,3,5-tetrahydro-4,1-benzoxazepin-2-one

  • MDL number MFCD32065925
  • Cas Number : 220107-63-1
  • Molecular Weight : 231.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-(trifluoromethyl)-1,5-dihydrobenzo[e][1,4]oxazepin-2(3H)-one
Inchi Code 1S/C10H8F3NO2/c11-10(12,13)7-1-2-8-6(3-7)4-16-5-9(15)14-8/h1-3H,4-5H2,(H,14,15)
Inchi Key DYDPYROSAJUOED-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430968
Availability details for
Quantity
  • Availability
  • Add to Cart