ENAH5802E04F Enamine

2-fluoro-2-(piperidin-3-ylidene)ethan-1-ol; trifluoroacetic acid

  • Cas Number : 1160945-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430960
Availability details for
Quantity
  • Availability
  • Add to Cart