ENAH5802E052 Enamine

6-chloro-N-(cyclopentylmethyl)pyridazine-3-carboxamide

  • MDL number MFCD12165868
  • Cas Number : 1179792-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435040
Availability details for
Quantity
  • Availability
  • Add to Cart