ENAH5802E053 Enamine

6-chloro-N-cyclohexylpyridazine-3-carboxamide

  • MDL number MFCD12165786
  • Cas Number : 1179087-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435038
Availability details for
Quantity
  • Availability
  • Add to Cart