ENAH5802E056 Enamine

6-chloro-N-(oxan-4-yl)pyridine-3-carboxamide

  • MDL number MFCD12088608
  • Cas Number : 1101793-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434389
Availability details for
Quantity
  • Availability
  • Add to Cart