ENAH5802E057 Enamine

8-bromo-1,2,3,5-tetrahydro-4,1-benzoxazepin-2-one

  • MDL number MFCD32068948
  • Cas Number : 2375273-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7436838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436838
Availability details for
Quantity
  • Availability
  • Add to Cart