ENAH5802E05C Enamine

7-bromo-3,3-dimethyl-1H,2H,3H,4H-pyrrolo[1,2-a]pyrazin-1-one

  • MDL number MFCD32068680
  • Cas Number : 1851197-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3071624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3071624
Availability details for
Quantity
  • Availability
  • Add to Cart