ENAH5802E05E Enamine

2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)-N-methylacetamide

  • MDL number MFCD16664863
  • Cas Number : 1205135-49-4
  • Molecular Weight : 246.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)-N-methylacetamide
Inchi Code 1S/C8H12BrN3O/c1-5-8(9)6(2)12(11-5)4-7(13)10-3/h4H2,1-3H3,(H,10,13)
Inchi Key YTBIXYVZGYXUHU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7437040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437040
Availability details for
Quantity
  • Availability
  • Add to Cart