ENAH5802E061 Enamine

7-bromo-3,3-difluoro-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD22547906
  • Cas Number : 1360928-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354943
Availability details for
Quantity
  • Availability
  • Add to Cart