ENAH5802E066 Enamine

6-chloro-N-(3,3,3-trifluoropropyl)pyridazine-3-carboxamide

  • MDL number MFCD18298266
  • Cas Number : 1342411-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433550
Availability details for
Quantity
  • Availability
  • Add to Cart