ENAH5802E067 Enamine

2-[2-(3-chlorophenyl)-2-oxoethylidene]-1,3-thiazolidin-4-one

  • Cas Number : 792954-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11225
Availability details for
Quantity
  • Availability
  • Add to Cart