ENAH5802E068 Enamine

N-(1H-pyrazol-4-yl)-2-(trifluoromethyl)benzamide

  • MDL number MFCD12075708
  • Cas Number : 1153804-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432271
Availability details for
Quantity
  • Availability
  • Add to Cart