ENAH5802E06B Enamine

N-[2-chloro-4-(trifluoromethyl)phenyl]-2-cyanoacetamide

  • MDL number MFCD18624328
  • Cas Number : 1519803-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439983
Availability details for
Quantity
  • Availability
  • Add to Cart