ENAH5802E071 Enamine

4-bromo-7-methoxyquinoline-6-carboxamide

  • MDL number MFCD30528197
  • Cas Number : 2089310-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442150
Availability details for
Quantity
  • Availability
  • Add to Cart