ENAH5802E076 Enamine

2-{5H,6H,7H-indeno[5,6-b]furan-3-yl}-N-(5-methyl-1,3-thiazol-2-yl)acetamide

  • Cas Number : 790245-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-252755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252755
Availability details for
Quantity
  • Availability
  • Add to Cart