ENAH5802E07D Enamine

5-bromo-N-[1-(4-ethoxyphenyl)ethyl]-2-methylbenzamide

  • Cas Number : 2375274-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7442400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7442400
Availability details for
Quantity
  • Availability
  • Add to Cart