ENAH5802E094 Enamine

3-hydroxy-3-methylbutanimidamide hydrochloride

  • Cas Number : 912578-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7441426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441426
Availability details for
Quantity
  • Availability
  • Add to Cart