ENAH5802E098 Enamine

methyl 5-iminopyrrolidine-3-carboxylate hydrochloride

  • MDL number MFCD32066042
  • Cas Number : 2361635-93-8
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C6H10N2O2.ClH/c1-10-6(9)4-2-5(7)8-3-4;/h2,4,8H,3,7H2,1H3;1H
Inchi Key LBIANUMFOHZLCX-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432328

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7432328
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart