ENAH5802E09B Enamine

6-methoxypyridine-2-carboximidamide hydrochloride

  • MDL number MFCD12755552
  • Cas Number : 1179361-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431145
Availability details for
Quantity
  • Availability
  • Add to Cart