ENAH5802E09D Enamine

5-methoxypyrazine-2-carboximidamide dihydrochloride

  • MDL number MFCD32068768
  • Cas Number : 2361732-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435374
Availability details for
Quantity
  • Availability
  • Add to Cart