ENAH5802E09E Enamine

6-fluoro-2-methyl-2,3-dihydro-1H-isoindol-1-imine hydrobromide

  • Cas Number : 2344679-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426016
Availability details for
Quantity
  • Availability
  • Add to Cart