ENAH5802E0A0 Enamine

4-ethenylbenzamide

  • MDL number MFCD18824576
  • Cas Number : 3661-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-170
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6772491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6772491
Availability details for
Quantity
  • Availability
  • Add to Cart