ENAH5802E0A1 Enamine

tert-butyl 5-amino-1,2,3,6-tetrahydropyrazine-1-carboxylate hydrochloride

  • MDL number MFCD32065836
  • Cas Number : 179686-22-7
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-amino-3,4-dihydropyrazine-1(2H)-carboxylate hydrochloride
Inchi Code 1S/C9H17N3O2.ClH/c1-9(2,3)14-8(13)12-5-4-11-7(10)6-12;/h6,11H,4-5,10H2,1-3H3;1H
Inchi Key CVXDBGBEEZXMAP-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-176383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176383
Availability details for
Quantity
  • Availability
  • Add to Cart