ENAH5802E0A2 Enamine

6-bromopyridine-2-carboximidamide hydrobromide

  • Cas Number : 2344679-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7424890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7424890
Availability details for
Quantity
  • Availability
  • Add to Cart