ENAH5802E0A3 Enamine

2-[4-(trifluoromethyl)phenyl]ethanimidamide hydrochloride

  • MDL number MFCD12922941
  • Cas Number : 1187436-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426134
Availability details for
Quantity
  • Availability
  • Add to Cart