ENAH5802E0A4 Enamine

1-[(4-methoxyphenyl)methyl]-1H-pyrazole-4-carboximidamide hydrochloride

  • MDL number MFCD31924531
  • Cas Number : 2344680-07-3
  • Molecular Weight : 266.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426822
Availability details for
Quantity
  • Availability
  • Add to Cart