ENAH5802E0A8 Enamine

N-methyl-N'-[1-(propan-2-yl)-1H-pyrazol-5-yl]thiophene-2-carboximidamide

  • Cas Number : 1006482-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14632
Availability details for
Quantity
  • Availability
  • Add to Cart